Polystyrene roller bottle with easy grip cap, Corning®
Lieferant: Corning
431644
734-2492EA 662 EUR
734-2492
Polystyrene roller bottle with easy grip cap, Corning®
Kolben Zellkulturkolben
The bottle is made with one piece seamless construction, has a wide mouth and printed graduations.
The bottle is manufactured from virgin polystyrene and the cap is made of high density polyethylene. Both resins meet the USP Class VI requirements for plastic containers and closures.
This product is sterilised by gamma radiation and is certified nonpyrogenic. It has printed lot numbers to aid in product traceability.
To be used with USP Class VI certified C-FLEX® tubing set.
The orange easy grip cap features large knurls designed for ergonomic handling. The bottle is not tissue-culture treated to minimise cell retention during seeding.