Multiflash
Lieferant: STANHOPE SETA
34200-0 34400-0
751-0070EA 0 EUR
751-0070 751-0072
Multiflash
Flammpunktprüfer
A fully automated instrument designed for testing the flashpoint of fuels, lubricating oils, solvents chemicals, waste materials and other substances.
  • Strict conformity to standard test methods
  • Gas or electric ignition, barometric correction
  • Displayed results in degrees Celsius or Fahrenheit
The instrument comprises a universal base unit that houses the display, control and data acquisition functions and a series of interchangeable modules that allow the measurement of the flash point following a range of different standard methods including ASTM, IP, DIN ISO etc. Automatic barometric correction.
Bestellinformation: Universal base unit is required for operation of the modules. The Cooling Boost module is required for operation of the Abel and TAg closed cup modules.
Lieferumfang: Supplied with DIPS pod, test area, igniter, appropriate detector, sample PRT and bath PRTs (if appropriate), interconnection leads and instruction manual.