Animal cell model
Lieferant: Binhold, Paul Lehrmittel
1000523
761-0465EA 794 EUR
761-0465
Animal cell model
Modelle Zoologie
The two piece animal cell model shows the form and structure of a typical animal cell as viewed by an electron microscope.
For purposes of better illustration, all important organelles are shown in raised relief and displayed in colour:
Nucleus, mitochondrion, smooth endoplasmic reticulum (ER), rough endoplasmic reticulum (ER), basal membrane, collagen fibres, Golgi apparatus, microvilli, lysosome.