Head with muscles, half
Lieferant: 3B Scientific
1000221
763-0100EA 833 EUR
763-0100
Head with muscles, half
Modelle Humanbiologie
The model shows all external structures such as blood vessels, nerves, etc. The median section shows all the structures of the head and neck. Dimensions: 22×18×46 cm.