picoLUCENT™ PLUS-AP kit
Lieferant: G Bioscienes
786-09AP 786-09APT
786-09APEA 508 EUR
786-09AP 786-09APT
picoLUCENT™ PLUS-AP kit
Elektrophorese-Reagenzien Western Blotting Reagents
The picoLUCENT™ PLUS-HRP kit is based on an ultra sensitive luminol substrate that produces chemiluminescence upon reaction with alkaline phosphatase. The chemiluminescence light emission can be recorded by a short exposure to autoradiography films.
  • Low picogram detection (10⁻¹²) of antigens
  • Low noise (signal/background ratio)
Lieferumfang: Primary antibody not included.