picoLUCENT™ PLUS-AP kit
Lieferant: G Bioscienes
786-09AP
786-09APT
786-09APEA
508
EUR
786-09AP
786-09APT
picoLUCENT™ PLUS-AP kit
Elektrophorese-Reagenzien
Western Blotting Reagents
The picoLUCENT™ PLUS-HRP kit is based on an ultra sensitive luminol substrate that produces chemiluminescence upon reaction with alkaline phosphatase. The chemiluminescence light emission can be recorded by a short exposure to autoradiography films.
- Low picogram detection (10⁻¹²) of antigens
- Low noise (signal/background ratio)
Lieferumfang: Primary antibody not included.