InGel™ for protein digestion
Lieferant: G Bioscienes
InGel™ provides a reliable method for the proteolytic digestion and subsequent extration of silver stained proteins in gel for subsequent analysis by mass spectrometry.
The protein spots are first excised from the gel and transferred to a proteomic grade tube. Silver stained gel pieces are washed with SilverOUT™ to remove inhibitory silver ions and then the gel pieces are treated with a mixture of our mass spectrometry grade trypsin and an optimal digestion buffer, which ensures reliable and efficient protein digestion.
The digested peptides are extracted with Pep-Extract™, a high diffusion peptide extraction buffer. The extracted peptides are suitable for mass spectrometry analysis without any subsequent treatments or cleaning procedures.
Lieferumfang: The InGel™ kit is supplied with all the necessary reagents for 100 protein spots and includes: SilverOUT™ destaining reagent; MSG-Trypsin™, a mass spectrometry grade trypsin with minimal autolysis and increases stability; trypsin digestion buffer, for optimal trypsin activity; and Pep-Extract™ Buffer, for high level peptide extraction.