FOCUS™ Yeast proteome kits
Lieferant: G Bioscienes
786-257
786-257EA
802
EUR
786-257
FOCUS™ Yeast proteome kits
Protein-Extraktion
Protein Extraction Kits
Specifically designed for yeast research and supplied with yeast specific reagents. Extracts and solubilises nearly all of the proteins from yeast, including membrane and soluble proteins. Extraction is based on gentle lysis of yeast cells with a yeast lytic enzyme preparation, LongLife™ Zymolyase®, which has improved stability and shelf life. Enzymatic action is followed by extraction of total proteome with the supplied strong chaotropic extraction buffer that solubilises even the most difficult proteins.
- Simple extraction protocol
- Supplied with yeast specific lytic enzyme preparation
- Supplied with a strong proprietary chaotropic extraction buffer
- Suitable for 50×60 μl yeast cell pellet preparations
- Suitable for sample preparation for 2D gel electrophoresis and other applications