Proteome kits, FOCUS™ Plant Proteome
Lieferant: G Bioscienes
786-259
786-259EA 782 EUR
786-259
Proteome kits, FOCUS™ Plant Proteome
Protein-Extraktion Protein Extraction Kits
Specifically designed for plant research and supplied with plant specific reagents, including reagents for removal of plant pigments and other natural products that may interfere with protein analysis. Extracts and solubilises nearly all of the proteins from plants, including membrane as well as soluble proteins. Supplied with a strong proprietary chaotropic extraction buffer to solubilise even the most difficult proteins.
  • Simple extraction protocol
  • Supplied with reagents for removal of plant pigments and other natural products that may interfere with protein analysis
  • Supplied with a proprietary chaotropic extraction buffer
  • Extracts plant proteome from 25×0,5 g plant tissue preparations
  • Suitable for sample preparation for 2D gel electrophoresis and other applications