High molecular weight transfer buffer (5X)
Lieferant: G Bioscienes
786-423
786-423EA
191
EUR
786-423
High molecular weight transfer buffer (5X)
Elektrophorese-Reagenzien
Western Blotting Reagents
High molecular weight transfer buffer (5X) is designed to facilitate the transfer of notoriously difficult high molecular weight proteins (>70 kDa) during Western blotting.