Biotin resin
Lieferant: G Bioscienes
786-598
786-598EA
808
EUR
786-598
Biotin resin
Protein-Extraktion
Proteinextraktions-Reagenzien
Immobilised biotin resin is designed for the high affinity chromatography purifications of avidin, streptavidin and neutravidin protein. The resin consists of biotin coupled to 6% cross-linked agarose.
- Strong affinity for avidin, streptavidin and neutravidin
- Reusable resin, at least 10 times
- Covalently coupled to limit leaching