Immobilised Iminobiotin Resin
Lieferant: G Bioscienes
786-599
786-599EA 446 EUR
786-599
Immobilised Iminobiotin Resin
Protein-Extraktion Proteinextraktions-Reagenzien
Immobilised Iminobiotin Resin consists of iminobiotin, a cyclic guanido analog of biotin, covalently coupled to 6% crosslinked agarose. The resin allows for the purification of avidin, streptavidin and Neutravidin and their subsequent gentle elution using non denaturing elution buffers.
  • Biotin binding capacity: 2 mg avidin/ml resin
  • No requirement for strong, denaturing elution buffers
  • Elutes at pH 4,0