Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE)

Lieferant: AAT BIOQUEST
100321L0 100321L1 100321L2
AATB100321L0EA 271 EUR
AATB100321L0 AATB100321L1 AATB100321L2
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE)
Antikörper
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR