Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (APC (Allophycocyanin))

Lieferant: AAT BIOQUEST
100411C0 100411C1 100411C2
AATB100411C0EA 271 EUR
AATB100411C0 AATB100411C1 AATB100411C2
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (APC (Allophycocyanin))
Antikörper
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR