Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PerCP/Cy5.5)

Lieferant: AAT BIOQUEST
101611U0 101611U1 101611U2
AATB101611U0EA 319 EUR
AATB101611U0 AATB101611U1 AATB101611U2
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (PerCP/Cy5.5)
Antikörper
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: PerCP/Cy5.5
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR