Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 790)

Lieferant: AAT BIOQUEST
101910M0 101910M1
AATB101910M0EA 371 EUR
AATB101910M0 AATB101910M1
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (iFluor® 790)
Antikörper
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 790
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR