Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (APC (Allophycocyanin)/iFluor® 700)
Lieferant: AAT BIOQUEST
10271110
AATB10271110EA 371 EUR
AATB10271110
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (APC (Allophycocyanin)/iFluor® 700)
Antikörper
O323 is an anti-human monoclonal antibody that is specific for the CD27 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: APC (Allophycocyanin)/iFluor® 700
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human