Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 800)

Lieferant: AAT BIOQUEST
103400N0 103400N1
AATB103400N0EA 371 EUR
AATB103400N0 AATB103400N1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 800)
Antikörper
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® 800
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR