Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 660)

Lieferant: AAT BIOQUEST
103800G0 103800G1
AATB103800G0EA 371 EUR
AATB103800G0 AATB103800G1
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 660)
Antikörper
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 660
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR