Anti-Human CD123 Mouse Monoclonal Antibody [clone: 12H7] (iFluor® 810)

Lieferant: AAT BIOQUEST
112310O0
AATB112310O0EA 373 EUR
AATB112310O0
Anti-Human CD123 Mouse Monoclonal Antibody [clone: 12H7] (iFluor® 810)
Antikörper
The 12H7 monoclonal antibody binds to human CD123, a 70 kD single-pass type I membrane protein often located on the surface of macrophages, hematopoietic progenitors and dendritic cells. CD123 acts in vital cellular pathways, for instance, the cytokine-mediated signaling pathway. From a research standpoint, it is of biological interest due to its association with essential macromolecules/ligands like CD131.

CD123 is a fairly uncommon antibody target, with a little more than 3900 publications in the last decade. Even still, CD123 is essential for immunology research, typically serving as aMore Product Information
Neuer Artikel bei VWR!
Bestellen

TECHNISCHE DATEN

Antigen-Name Human CD123
Environmentally Preferable
Antigen-Synonyme IL-3Rα
Klonalität Monoklonal
Konjugation iFluor® 810
Klon 12H7
Reaktivität Human
Wirt Mouse
Isotyp IgG1 kappa
ImmunFluoreszenz Yes
Durchflusszytometrie Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR