Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104] (mFluor™ Violet 510)

Lieferant: AAT BIOQUEST
12000110
AATB12000110EA 371 EUR
AATB12000110
Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104] (mFluor™ Violet 510)
Antikörper
OX-104 is an anti-human monoclonal antibody that forms an immune complex with the CD200 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: OX-104
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR