Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 594)

Lieferant: AAT BIOQUEST
16404
AATB16404EA 230 EUR
AATB16404
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 594)
Antikörper
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).

  • Affinity-purified
  • Bright signal
  • Cross-adsorbed

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR