Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 546)

Lieferant: AAT BIOQUEST
16457
AATB16457EA 120 EUR
AATB16457
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 546)
Antikörper
iFluor® 546 is a bright orange fluorescent dye.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Bitte beachten For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 546
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR