Anti-IgG Mouse Polyclonal Antibody (trFluor™ Tb)

Lieferant: AAT BIOQUEST
16756
AATB16756EA 3050 EUR
AATB16756
Anti-IgG Mouse Polyclonal Antibody (trFluor™ Tb)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: trFluor™ Tb
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR