Tetracysteine FLASH Reagent *2 mM DMSO Solution*
Lieferant: AAT BIOQUEST
22330
AATB22330EA 568 EUR
AATB22330
Tetracysteine FLASH Reagent *2 mM DMSO Solution*
Tests Proteintests
FLASH is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
Bitte beachten For research use only.