Tetracysteine REASH Reagent *2 mM DMSO Solution*
Lieferant: AAT BIOQUEST
22332
AATB22332EA
568
EUR
AATB22332
Tetracysteine REASH Reagent *2 mM DMSO Solution*
Tests
Proteintests
REASH is a resorufin derivative, modified to contain two arsenic atoms at a set distance from each other.
- Resorufin derivative
- High affinity for thiols
- Multiple applications
Bitte beachten For research use only.