Tetracysteine REASH Acetate *2 mM DMSO Solution*
Lieferant: AAT BIOQUEST
REASH acetate is a cell-permeable version of REASH, which is a resorufin derivative, modified to contain two arsenic atoms at a set distance from each other.
- Fluorescein derivative
- High affinity for thiols
- Multiple applications
Bitte beachten For research use only.