AAT Bioquest's anti-human secondary antibodies have well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
- Purified and quality-tested for consistent results
- Suitable for multiple applications
- Purified through affinity chromatography
Multiple secondary antibodies can bind to a single primary antibody.
Bitte beachten Research use only.
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: IgG