Amplite® Fluorimetric Biotin Quantitation Kit
Lieferant: AAT BIOQUEST
5538
AATB5538EA 309 EUR
AATB5538
Amplite® Fluorimetric Biotin Quantitation Kit
Tests Proteintests
Designed for consistent and dependable results. Amplite® Fluorimetric Biotin Quantitation Kit uses Biotinylite™ Green, a fluorogenic biotin sensor.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
This fluorescence-based assay is much more sensitive than the commonly used colorimetric HABA assay.
Lieferumfang: Shipped with blue ice.
Bitte beachten Research use only.