Anti-HLCS Rabbit Polyclonal Antibody

Lieferant: Abcam
AB100925-100
ABCAAB100925-100EA 774 EUR
ABCAAB100925-100
Anti-HLCS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HLCS (holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase (ATP-hydrolysing)) ligase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR