Anti-UBA52 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB104455-200
ABCAAB104455-200EA 700 EUR
ABCAAB104455-200
Anti-UBA52 Rabbit Polyclonal Antibody
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: UBA52 (ubiquitin A-52 residue ribosomal protein fusion product 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR