Anti-Glutamate Transporter 1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB106289-200
ABCAAB106289-200EA 918 EUR
ABCAAB106289-200
Anti-Glutamate Transporter 1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Glutamate Transporter 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR