Anti-Epoxide hydrolase Mouse Monoclonal Antibody [clone: 15B2AD9]

Lieferant: Abcam
AB110307-100
ABCAAB110307-100EA 669 EUR
ABCAAB110307-100
Anti-Epoxide hydrolase Mouse Monoclonal Antibody [clone: 15B2AD9]
Antikörper
Mouse monoclonal [15B2AD9] to Epoxide hydrolase.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Epoxide hydrolase
Clonality: Monoclonal
Clone: 15B2AD9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR