Anti-PTP zeta/ Phosphacan Rabbit Polyclonal Antibody

Lieferant: Abcam
AB110479-100
ABCAAB110479-100EA 638 EUR
ABCAAB110479-100
Anti-PTP zeta/ Phosphacan Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal to PTP zeta/ Phosphacan.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PTP zeta/ Phosphacan
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR