Anti-Nicotinic Acetylcholine Receptor alpha 7 Goat Polyclonal Antibody

Lieferant: Abcam
AB110851-50
ABCAAB110851-50EA 918 EUR
ABCAAB110851-50
Anti-Nicotinic Acetylcholine Receptor alpha 7 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nicotinic Acetylcholine Receptor alpha 7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR