Anti-14-3-3 beta/zeta Rabbit Polyclonal Antibody

Lieferant: Abcam
AB111856-100
ABCAAB111856-100EA 638 EUR
ABCAAB111856-100
Anti-14-3-3 beta/zeta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 14-3-3 beta/zeta (14-3-3 beta/zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR