Anti-IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abcam
AB112833-500
ABCAAB112833-500EA 412 EUR
ABCAAB112833-500
Anti-IgM Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Secondary
Antigen: IgM (Dog) (dog IgM)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR