Anti-Cytokeratin Mouse Monoclonal Antibody [clone: CK-cocktail]

Lieferant: Abcam
AB115959-1000
ABCAAB115959-1000EA 808 EUR
ABCAAB115959-1000
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: CK-cocktail]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: Cytokeratin
Clonality: monoclonal
Clone: CK-cocktail
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR