Anti-SIRP alpha Rabbit Polyclonal Antibody

Lieferant: Abcam
AB116254-50
ABCAAB116254-50EA 918 EUR
ABCAAB116254-50
Anti-SIRP alpha Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SIRP alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR