Anti-RBBP9 Mouse Monoclonal Antibody [clone: 4E1]

Lieferant: Abcam
AB118023-100
ABCAAB118023-100EA 825 EUR
ABCAAB118023-100
Anti-RBBP9 Mouse Monoclonal Antibody [clone: 4E1]
Antikörper
Type: Primary
Antigen: RBBP9 (retinoblastoma binding protein 9)
Clonality: Monoclonal
Clone: 4E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR