Anti-HLA DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: LN3]

Lieferant: Abcam
AB1182-500
ABCAAB1182-500EA 882 EUR
ABCAAB1182-500
Anti-HLA DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: LN3]
Antikörper
Type: Primary
Antigen: HLA DR
Clonality: monoclonal
Clone: LN3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR