Anti-HLA-DR Rabbit Polyclonal Antibody

Lieferant: Abcam
AB118347-100
ABCAAB118347-100EA 931 EUR
ABCAAB118347-100
Anti-HLA-DR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HLA-DR
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR