Anti-CD3 zeta Armenian Hamster Monoclonal Antibody [clone: H146-968]

Lieferant: Abcam
AB119827-100
ABCAAB119827-100EA 1000 EUR
ABCAAB119827-100
Anti-CD3 zeta Armenian Hamster Monoclonal Antibody [clone: H146-968]
Antikörper
Bitte beachten

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: CD3 zeta
Clonality: monoclonal
Clone: H146-968
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR