Anti-FYN Mouse Monoclonal Antibody [clone: 2A10]

Lieferant: Abcam
AB119855-100
ABCAAB119855-100EA 825 EUR
ABCAAB119855-100
Anti-FYN Mouse Monoclonal Antibody [clone: 2A10]
Antikörper
Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR