Anti-GABA Transporter 3 / GAT 3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB122430-100
ABCAAB122430-100EA 912 EUR
ABCAAB122430-100
Anti-GABA Transporter 3 / GAT 3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GABA Transporter 3 / GAT 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR