Anti-DDDDK tag (Binds to FLAG® tag sequence) Goat Polyclonal Antibody (Agarose)

Lieferant: Abcam
AB1240-100
ABCAAB1240-100EA 688 EUR
ABCAAB1240-100
Anti-DDDDK tag (Binds to FLAG® tag sequence) Goat Polyclonal Antibody (Agarose)
Antikörper
Agarose Goat polyclonal to DDDDK tag (Binds to FLAG® tag sequence).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: DDDDK tag (Binds to FLAG® tag sequence)
Clonality: Polyclonal
Clone:
Conjugation: Agarose
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR