Anti-HLA-DR Rabbit Monoclonal Antibody [clone: EPR3691]

Lieferant: Abcam
AB124809-100
ABCAAB124809-100EA 928 EUR
ABCAAB124809-100
Anti-HLA-DR Rabbit Monoclonal Antibody [clone: EPR3691]
Antikörper
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone: EPR3691
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR