Anti-DDDDK tag (Binds to FLAG® tag sequence) Mouse Monoclonal Antibody [clone: FG4R]

Lieferant: Abcam
AB125243-100
ABCAAB125243-100EA 475 EUR
ABCAAB125243-100
Anti-DDDDK tag (Binds to FLAG® tag sequence) Mouse Monoclonal Antibody [clone: FG4R]
Antikörper
Mouse monoclonal [FG4R] to DDDDK tag (Binds to FLAG® tag sequence).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: DDDDK tag (Binds to FLAG® tag sequence)
Clonality: Monoclonal
Clone: FG4R
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR