Anti-SMC1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB1276-50
ABCAAB1276-50EA 963 EUR
ABCAAB1276-50
Anti-SMC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMC1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR