Anti-Serotonin transporter Goat Polyclonal Antibody

Lieferant: Abcam
AB130130-200
ABCAAB130130-200EA 918 EUR
ABCAAB130130-200
Anti-Serotonin transporter Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Serotonin transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR