Anti-Dopamine Transporter Goat Polyclonal Antibody

Lieferant: Abcam
AB130140-200
ABCAAB130140-200EA 825 EUR
ABCAAB130140-200
Anti-Dopamine Transporter Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Dopamine Transporter
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR